ENA245485996 Enamine

[1-(1-benzofuran-2-yl)ethyl](propyl)amine hydrochloride

  • MDL number MFCD13195904
  • Cas Number : 1197742-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53569

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53569
Availability details for
Quantity
  • Availability
  • Add to Cart