ENA245486005 Enamine

methyl[(2-methyl-1,3-thiazol-4-yl)methyl]amine

  • MDL number MFCD06738964
  • Cas Number : 144163-81-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27495

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27495
Availability details for
Quantity
  • Availability
  • Add to Cart