ENA245486006 Enamine

3-(5-methyl-2-oxo-2,3-dihydro-1H-imidazol-1-yl)benzonitrile

  • MDL number MFCD13195909
  • Cas Number : 1197714-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53589
Availability details for
Quantity
  • Availability
  • Add to Cart