ENA245486024 Enamine

3-[2-(4-chlorophenoxy)-2-methylpropanoyl]-1,3-thiazolidine-4-carboxylic acid

  • MDL number MFCD00869162
  • Cas Number : 64179-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53608
Availability details for
Quantity
  • Availability
  • Add to Cart