ENA245486041 Enamine

5-(2,4-dimethylphenyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD02664091
  • Cas Number : 183882-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53639
Availability details for
Quantity
  • Availability
  • Add to Cart