ENA245486045 Enamine

5-(3,4-difluorophenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09804932
  • Cas Number : 1016711-54-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-53643

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53643
Availability details for
Quantity
  • Availability
  • Add to Cart