ENA245486046 Enamine

hexanethioamide

  • MDL number MFCD09810087
  • Cas Number : 16525-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53644
Availability details for
Quantity
  • Availability
  • Add to Cart