ENA245486050 Enamine

1-(3,5-difluorophenyl)-2-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD09806122
  • Cas Number : 1016509-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53648

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53648
Availability details for
Quantity
  • Availability
  • Add to Cart