ENA245486055 Enamine

3-(chloromethyl)-5-(propan-2-yl)-1,2,4-oxadiazole

  • MDL number MFCD09864313
  • Cas Number : 189130-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-53654

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53654
Availability details for
Quantity
  • Availability
  • Add to Cart