ENA245486056 Enamine

4-[4-(but-2-en-1-yloxy)phenyl]butan-2-one

  • Cas Number : 1807920-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53655
Availability details for
Quantity
  • Availability
  • Add to Cart