ENA245486059 Enamine

2-(2,5-dimethyl-1H-pyrrol-1-yl)propanoic acid

  • MDL number MFCD02302091
  • Cas Number : 130016-65-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53659
Availability details for
Quantity
  • Availability
  • Add to Cart