ENA245486071 Enamine

3-(3-amino-4-fluorophenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD11167822
  • Cas Number : 1042623-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53681

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53681
Availability details for
Quantity
  • Availability
  • Add to Cart