ENA245486080 Enamine

N-[(4-chlorophenyl)methyl]aniline

  • MDL number MFCD03210840
  • Cas Number : 4750-61-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53690
Availability details for
Quantity
  • Availability
  • Add to Cart