ENA245486210 Enamine

1-(4-ethylphenoxy)-3-(methylamino)propan-2-ol hydrochloride

  • MDL number MFCD13195939
  • Cas Number : 1209814-95-8
  • Molecular Weight : 245.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53786
Availability details for
Quantity
  • Availability
  • Add to Cart