ENA245486231 Enamine

5-chloro-2-(4-methylpiperazin-1-yl)aniline dihydrochloride

  • MDL number MFCD13195988
  • Cas Number : 1210682-18-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54033
Availability details for
Quantity
  • Availability
  • Add to Cart