ENA245486245 Enamine

1-benzyl-3-methyl-1H-pyrazolo[3,4-b]pyridin-5-amine

  • MDL number MFCD13196032
  • Cas Number : 1211409-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54276
Availability details for
Quantity
  • Availability
  • Add to Cart