ENA245486247 Enamine

1-[(4-hydrazinyl-3-nitrobenzene)sulfonyl]pyrrolidine hydrochloride

  • MDL number MFCD13196037
  • Cas Number : 1210618-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54329

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54329
Availability details for
Quantity
  • Availability
  • Add to Cart