ENA245486254 Enamine

(1R)-1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethan-1-amine hydrochloride

  • MDL number MFCD13196051
  • Cas Number : 1212291-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54373

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54373
Availability details for
Quantity
  • Availability
  • Add to Cart