ENA245486260 Enamine

2-cyclopropylpropan-1-amine hydrochloride

  • MDL number MFCD13196072
  • Cas Number : 1209286-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146...148
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54480

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54480
Availability details for
Quantity
  • Availability
  • Add to Cart