ENA245486264 Enamine

4-(2-chloropropanoyl)phenyl 2-chloropropanoate

  • MDL number MFCD13196076
  • Cas Number : 1208654-68-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55...57
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54494

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54494
Availability details for
Quantity
  • Availability
  • Add to Cart