ENA245486265 Enamine

4,4,4-trifluoro-3-hydroxy-3-(1H-imidazol-2-yl)butanoic acid

  • MDL number MFCD13196079
  • Cas Number : 1211096-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54498

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54498
Availability details for
Quantity
  • Availability
  • Add to Cart