ENA245486266 Enamine

4-bromo-2-(dimethylamino)benzoic acid hydrochloride

  • MDL number MFCD13196080
  • Cas Number : 1209966-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-54499

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54499
Availability details for
Quantity
  • Availability
  • Add to Cart