ENA245486267 Enamine

2-(2-methyl-1H-imidazol-1-yl)aniline dihydrochloride

  • MDL number MFCD13196083
  • Cas Number : 1210033-60-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54504

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54504
Availability details for
Quantity
  • Availability
  • Add to Cart