ENA245486272 Enamine

cyclooctylmethanamine hydrochloride

  • MDL number MFCD13196090
  • Cas Number : 1209904-16-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54583

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54583
Availability details for
Quantity
  • Availability
  • Add to Cart