ENA245486288 Enamine

7-(difluoromethoxy)-1,2,3,4-tetrahydroisoquinoline

  • MDL number MFCD13196121
  • Cas Number : 1209300-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54721
Availability details for
Quantity
  • Availability
  • Add to Cart