ENA245486290 Enamine

5-(chloromethyl)-3-(3-methylbutyl)-1,2,4-oxadiazole

  • MDL number MFCD13196129
  • Cas Number : 1209200-59-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54770

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54770
Availability details for
Quantity
  • Availability
  • Add to Cart