ENA245486293 Enamine

[4-(4-chlorophenyl)-1,3-thiazol-2-yl]methanamine hydrochloride

  • MDL number MFCD13196144
  • Cas Number : 1208832-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-54845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54845
Availability details for
Quantity
  • Availability
  • Add to Cart