ENA245486294 Enamine

7-({3-[(ethoxycarbonyl)amino]phenoxy}sulfonyl)-2,3-dihydro-1,4-benzodioxine-5-carboxylic acid

  • MDL number MFCD13196147
  • Cas Number : 1209657-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54873

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54873
Availability details for
Quantity
  • Availability
  • Add to Cart