ENA245486297 Enamine

2-tert-butyl-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD13196151
  • Cas Number : 1211695-52-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54886

Safety Information

msds https://www.enaminestore.com/catalog/EN300-54886
Availability details for
Quantity
  • Availability
  • Add to Cart