ENA245486298 Enamine

3-phenyl-4H-1lambda6-pyrido[2,3-e][1,2,4]thiadiazine-1,1-dione

  • MDL number MFCD13196152
  • Cas Number : 216507-85-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-54900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-54900
Availability details for
Quantity
  • Availability
  • Add to Cart