ENA245486305 Enamine

6-methyl-2-(pyrrolidin-1-yl)pyridine-3-carbonitrile

  • MDL number MFCD13196167
  • Cas Number : 1209155-95-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-55057

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55057
Availability details for
Quantity
  • Availability
  • Add to Cart