ENA245486310 Enamine

N-benzyl-N-methyl-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD13196174
  • Cas Number : 1208892-07-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55091

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55091
Availability details for
Quantity
  • Availability
  • Add to Cart