ENA245486314 Enamine

6-acetyl-1,3,7-trimethyl-1H,2H,3H,4H,7H-pyrrolo[2,3-d]pyrimidine-2,4-dione

  • MDL number MFCD13196181
  • Cas Number : 119350-44-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55164
Availability details for
Quantity
  • Availability
  • Add to Cart