ENA245486321 Enamine

2-phenoxypyridine-4-carboximidamide hydrochloride

  • MDL number MFCD13196189
  • Cas Number : 82019-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55200

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-55200
Availability details for
Quantity
  • Availability
  • Add to Cart