ENA245486322 Enamine

methyl 2-(fluorosulfonyl)acetate

  • MDL number MFCD13196190
  • Cas Number : 41505-91-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-55203

Safety Information

msds https://www.enaminestore.com/catalog/EN300-55203
Availability details for
Quantity
  • Availability
  • Add to Cart