ENA245486326 Enamine

2-(2-amino-5-fluorophenoxy)ethan-1-ol

  • MDL number MFCD11147836
  • Cas Number : 1021022-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53715

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53715
Availability details for
Quantity
  • Availability
  • Add to Cart