ENA245486338 Enamine

2-fluoro-3,5-dinitrobenzoic acid

  • MDL number MFCD00185653
  • Cas Number : 445-65-8
  • Molecular Weight : 230.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 200-201
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53917

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P273,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-53917
pictograms GHS07
hstatements H302,H315,H319,H335,H413
Availability details for
Quantity
  • Availability
  • Add to Cart