ENA245486345 Enamine

6,8-difluoro-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD01764784
  • Cas Number : 259655-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53717
Availability details for
Quantity
  • Availability
  • Add to Cart