ENA245486347 Enamine

3-(1,3-dioxaindane-5-amido)thiophene-2-carboxylic acid

  • MDL number MFCD09050421
  • Cas Number : 926192-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53930

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53930
Availability details for
Quantity
  • Availability
  • Add to Cart