ENA245486349 Enamine

{2-hydroxy-3-[2-(propan-2-yloxy)ethoxy]propyl}(methyl)amine

  • MDL number MFCD11135173
  • Cas Number : 1019624-79-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53933
Availability details for
Quantity
  • Availability
  • Add to Cart