ENA245486350 Enamine

[3-(benzyloxy)-2-hydroxypropyl](methyl)amine

  • MDL number MFCD05668683
  • Cas Number : 57962-52-4
  • Molecular Weight : 195.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53934

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53934
Availability details for
Quantity
  • Availability
  • Add to Cart