ENA245486353 Enamine

{4-[3-(trifluoromethyl)phenyl]oxan-4-yl}methanamine

  • MDL number MFCD10690803
  • Cas Number : 1152568-50-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-53719

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53719
Availability details for
Quantity
  • Availability
  • Add to Cart