ENA245486354 Enamine

{3-ethoxy-2-[(5-methyl-1,2-oxazol-3-yl)methoxy]phenyl}methanol

  • MDL number MFCD12545506
  • Cas Number : 1099647-85-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53720
Availability details for
Quantity
  • Availability
  • Add to Cart