ENA245486355 Enamine

3-cyclopropyl-3-azabicyclo[3.3.1]nonan-9-one

  • MDL number MFCD13806402
  • Cas Number : 1000931-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53721
Availability details for
Quantity
  • Availability
  • Add to Cart