ENA245486367 Enamine

1-(4-fluorophenyl)-4-hydroxy-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD11133587
  • Cas Number : 1152602-19-1
  • Molecular Weight : 222.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-53955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53955
Availability details for
Quantity
  • Availability
  • Add to Cart