ENA245486378 Enamine

2-({[4-(benzyloxy)phenyl]methyl}amino)acetamide

  • MDL number MFCD09043791
  • Cas Number : 774506-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-53966

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53966
Availability details for
Quantity
  • Availability
  • Add to Cart