ENA245486383 Enamine

4-bromo-6-(chloromethyl)-1,3-dioxaindane

  • MDL number MFCD11182162
  • Cas Number : 65673-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53972
Availability details for
Quantity
  • Availability
  • Add to Cart