ENA245486387 Enamine

pentane-1,2-diamine

  • MDL number MFCD12190269
  • Cas Number : 52940-41-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53976

Safety Information

msds https://www.enaminestore.com/catalog/EN300-53976
Availability details for
Quantity
  • Availability
  • Add to Cart