ENA245486393 Enamine

1-(morpholine-4-carbonyl)piperidine-4-carboxylic acid

  • MDL number MFCD10012232
  • Cas Number : 1018574-31-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53988
Availability details for
Quantity
  • Availability
  • Add to Cart