ENA245486394 Enamine

4,4,4-trifluoro-3-hydroxy-3-(4-methyl-1,3-thiazol-2-yl)butanoic acid

  • MDL number MFCD13195976
  • Cas Number : 1197469-61-6
  • Molecular Weight : 255.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-53989

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-53989
Availability details for
Quantity
  • Availability
  • Add to Cart